Learn More
BD Vacutainer™ General Use Syringe Needles
Click to view available options
Needle Gauge:
18 G
Unit Size:
Pack of 1000
Description
Blunt fill needle provides a substitute for conventional needles
- Provides the convenience of pre-attached combos along with clinical flexibility
- Bridges medication preparation and administration; effective devices for filling and transferring medications from vials, ampoules and IV bags to their final medication delivery reservoirs
- Designed to minimize the risk of needlesticks; requires 10X more puncture penetration force than a conventional sharp
- Lower cost alternative than sharps safety engineered devices
- 18 gauge blunt needles allow for rapid transfer and aspiration without coring
- 1.5 in. can easily penetrate both single and double septum barriers
Specifications
Specifications
| Needle Gauge | 18 G |
| For Use With (Application) | Provides a substitute for conventional needles, general use |
| Length (English) Needle | 1.5 in. |
| Product Type | Syringe Needle |
| Sterility | Sterile |