missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD V500 Streptavidin
Second-step reagent for the indirect immunofluorescent staining of cells in combination with biotinylated primary antibodies for flow cytometric analysis
Brand: BD 561419
Additional Details : Weight : 0.01000kg
Description
- Streptavidin protein conjugated to BD Horizon V500
- Clone: Streptavidin
- Concentration: 0.5mg/mL
- Reactivity: QC Testing: Mouse; Tested in development: Human
Flow Cytometry
Specifications
0.5mg/mL | |
7.3 | |
4°C | |
Flow Cytometry |
Mouse | |
0.1mg | |
Solution | |
Mouse Streptavidin BD Horizon V500 |
For research use only.