missing translation for 'onlineSavingsMsg'
Learn More

BD Transduction Laboratories Mouse Liver Lysate

Product Code. 15865939
Click to view available options
Quantity:
500 μg
Unit Size:
Each
This item is not returnable. View return policy
Do not dilute or boil

Product Code. 15865939

Brand: BD 611458

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Do not dilute or boil

For use as a control for western blotting

  • This lysate can be a rich source of endogenously expressed proteins
  • Provided in a denatured format
  • Ready to use for immediate loading onto an SDS-PAGE gel

TRUSTED_SUSTAINABILITY

Specifications

Tissue Liver
Quantity 500 μg
Storage Buffer SDS-PAGE Gel
Host Species Mouse
Concentration 1 mg/mL
Content And Storage -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.