missing translation for 'onlineSavingsMsg'
Learn More

BD Stat Activation Sampler Kit

For blotting procedures

Brand:  BD 612477

Product Code. 15816199

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Alert:

Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing


Includes: Stat1 (C-terminus), Stat1 (pY701), Stat3, Stat3 (pY705), Stat5, Stat5 (pY694), Stat6, Stat6 (pY641)
This item is not returnable. View return policy

Description

Description

The Stat proteins function both as cytoplasmic signal transducers and as activators of transcription. These proteins can contain SH2 and SH3 domains and are activated by both tyrosine and serine phosphorylation.

  • Concentration: 250μg/mL
  • Format: Purified

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research use only