missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD SL-29 Cell Lysate
Click to view available options
Quantity:
500 μg
Unit Size:
1 set
Description
Includes
Chicken Embryo Fibroblast
- Useful control for the western blotting application
- Can be rich in endogenously expressed proteins
- Storage buffer: 1mg/mL in SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
Specifications
Specifications
| Tissue | Embryo Fibroblast |
| Quantity | 500 μg |
| Storage Buffer | 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol |
| Host Species | Chicken |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction