Learn More
BD Recombinant Soluble Human Fas
Description
Fas (APO-1, CD95) is an ∽42 kDa cell surface protein that mediates apoptosis when cross-linked with agonistic anti-Fas antibodies or Fas ligand (FasL). Fas belongs to the TNF (tumor necrosis factor)/NGF (nerve growth factor) receptor family, and is expressed in various tissue and cells including the thymus, liver, ovary and lung. FasL is a 40 kDa TNF family member membrane protein that induces apoptosis by binding to Fas, its cell-surface receptor. FasL is expressed on activated T and NK cells. Both Fas and FasL are thought to play an important role in the apoptotic processes that take place during T cell development. FasδTM has 63-base pair deletion starting at nucleotide sequence (700) GA TCC AGA and ending at nucleotide sequence GTT TGG G (762).FasδTM has a reduced molecular weight of ∽40 kDa.
- Alias: FasδTM
- Molecular weight: ∽40 kDa
- Storage buffer: Aqueous buffered solution containing BSA and glycerol
- Protein purity and integrity were verified by SDS-PAGE, and ELISA
Avoid multiple freeze-thaws of product.
Western blot, ELISA Standard, Functional assay
Specifications
Specifications
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | Aqueous Buffered solution containing BSA and Glycerol |
| Molecular Weight (g/mol) | 42 |
| Name | Fas (Soluble) Recombinant Human Protein |
| Preparation Method | Expressed in the Baculovirus expression vector system and purified from insect cell lysate using an anti-Fas Antibody |
| Purification Method | ELISA, SDS-PAGE |
| Quantity | 10 μg |
| Storage Requirements | -80°C |
| Recombinant | Recombinant |
| Form | Solution |
Research use only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.