missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Rat Interleukin-4 (IL-4)

Product Code. 15877998 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μ g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877998 5 μ g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15877998 Supplier BD Supplier No. 555107
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent lymphoid cell growth factor

  • Stimulate growth, differentiation, survivability, function of cells of hematopoietic and non-hematopoietic origin
  • Binds to specific high affinity receptors
  • Gene product is 17 -20 kD glycoprotein containing 123 amino acid residues
  • Storage buffer: frozen aqueous buffered solution containing BSA

ELISA standard, bioassay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
Quantity 5 μ g
Source Rat

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.