missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Rat IFN-γ

Product Code. 15836188
Click to view available options
:
Recombinant Rat IFN-gamma; 100μg/mL; 5μg
This item is not returnable. View return policy

Product Code. 15836188

Brand: BD 550072

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent multifunctional cytokine, secreted by activated NK cells and CD4+TCRαβ+, CD8+TCRαβ+, and TCRγδ+T cells

Supplied as a frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution, and containing 2mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives

  • Reavtivity: Rat
  • Storage buffer: Frozen aqueous buffered solution containing BSA
  • Activity Test: Biological activity is measured in an anti-viral bioassay using L929 indicator cells
  • Specific Activity Range: 0.1-1.0x108Unit/mg
  • ED50: 0.1 - 1.0 ng/ml
  • Observed dose response relationship: 100 fold

Bioassay

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100μg/mL
For Use With (Application) Bioassay
Formulation Frozen aqueous Buffered solution containing BSA
Name Rat IFN-gamma Recombinant Protein
Quantity 5μg
Storage Requirements -80°C
Cross Reactivity Rat
Recombinant Recombinant
Form Solution

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.