missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Mouse Interleukin-6 (IL-6)
Click to view available options
Quantity:
5 μg
Unit Size:
Each
Description
- 21.7kD multifunctional cytokine
- Contains 187 amino acid residues
- Targets hepatocytes, B and T lymphocytes, neurons, tumor cells and multipotent hematopoietic cells
- Storage buffer: frozen aqueous buffered solution containing BSA and glycerol
Bioassay, ELISA standard, blocking
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | Bioassay, Blocking Agent, ELISA Standard |
| Formulation | Frozen aqueous Buffered solution containing BSA and Glycerol |
| Molecular Weight (g/mol) | 21.7 |
| Name | Mouse IL-6 Recombinant Protein |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Mouse |
| Recombinant | Recombinant |
| Form | Solution |
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction