missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Mouse Interleukin-2 (IL-2)
Click to view available options
:
Recombinant Mouse IL-2; 200μg/mL; 20μg
Description
Quantitative standard for measuring mouse IL-2 protein levels in an IL-2 specific sandwich ELISA with the purified JES6-1A12 antibody as capture antibody and the biotinylated JES6-5H4 antibody as detection antibody
- Storage buffer: Frozen aqueous buffered solution containing BSA
- Biological Activity: Activity Range is measured in a proliferation assay using CTLL-2 indicator cells
- Specific Activity: 0.1 - 1.0 x 1099 Units™/mg
- ED50: 10 - 100 pg/ml
Bioassay, Enzyme-linked immunosorbent assay standard
Specifications
Specifications
| Concentration | 200μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard |
| Formulation | Frozen aqueous Buffered solution containing BSA |
| Molecular Weight (g/mol) | 17.2 |
| Name | Mouse IL-2 Recombinant Protein |
| Quantity | 20μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Mouse |
| Recombinant | Recombinant |
| Form | Solution |
Research Use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction