missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Mouse IFN-γ
Click to view available options
Quantity:
10 μg
Unit Size:
Each
Description
- Purified by immunoaffinity chromatography
Enzyme Linked Immunosorbent Assay, BioAssay
Specifications
Specifications
| Concentration | 200μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard |
| Name | Mouse IFN-gamma Recombinant Protein |
| Quantity | 10 μg |
| Storage Requirements | -80°C |
| Recombinant | Recombinant |
| Form | Liquid |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction