missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Human Monokine induced by γ interferon (MIG)

Product Code. 15837818
Click to view available options
Quantity:
5 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15837818

Brand: BD 554636

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pure determined by SDS-PAGE and an absorbance assay based on the Beers-Lambert law

Ffrozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution containing 10% glycerol and mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives

  • Reactivity: Human
  • Storage Buffer: Frozen aqueous buffered solution containing BSA

Enzyme-linked immunosorbent assay standard

TRUSTED_SUSTAINABILITY

Specifications

Formulation Frozen aqueous Buffered solution containing BSA
Name Human MIG Recombinant Protein
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
Form Solution

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.