missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Human Monocyte chemoattractant protein-1 (MCP-1)

Product Code. 15807818
Click to view available options
Quantity:
10 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15807818

missing translation for 'mfr': BD 554620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent chemoattractant for monocytes and it also activates lymphocytes, basophils and NK cells

Frozen liquid comprised of 0.22 ++m sterile-filtered aqueous buffered solution (pH 7.2) and containing 2 mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives

  • Reactivity: Human
  • Storage Buffer: Frozen aqueous buffered solution containing BSA

Enzyme-linked immunosorbent assay standard

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioassay, ELISA Standard
Formulation Frozen aqueous Buffered solution containing BSA
Name Human MCP-1 Recombinant Protein
Quantity 10 μg
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
Form Solution

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.