missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Human Interleukin-2 (IL-2)

Product Code. 15887798
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15887798 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15887798 Supplier BD Supplier No. 554603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purified by immunoaffinity chromatography and was found to be >95% pure by SDS-PAGE analysis, and an absorbance assay based on the Beers-Lambert law

Frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution, pH 7.2, and containing 2.0mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives

  • Reactivity: Human
  • Storage Buffer: Frozen aqueous buffered solution containing BSA and glycerol
  • Biological Activity; The assay range is measured in a proliferation assay using CTLL-2 indicator cells
  • Specific Activity: 0.06 - 1.0 x 109 Units/mg
  • ED50: 10 to 150pg/ml
  • Observed dose response: 50 fold

Enzyme-linked immunosorbent assay standard, Bioassay, intracellular block/flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Concentration 200μg/mL
For Use With (Application) Bioassay, ELISA Standard, Intracellular Block/Flow Cytometry
Formulation Frozen aqueous Buffered solution containing BSA and Glycerol
Name Human IL-2 Recombinant Protein
pH Range 7.2
Purification Method Immunoaffinity Chromatography, SDS-PAGE
Quantity 10 μg
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
Form Solution
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.