missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Human Interleukin-10 (IL-10)
Click to view available options
Quantity:
5 μg
Unit Size:
Each
Description
Frozen, sterile-filtered aqueous buffered solution containing glycerol and biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
- Reactivity: Human
- Bioassay: The activity range is measured in a proliferation assay using MC/9 indicator cells
- Specific Activity: 0.5 - 3.3 x 107 Units/mg
- ED50: 0.3 to 2.0ng/ml
- Observed dose response: >250 fold
Bioassay, enzyme-linked immunosorbent assay standard, intracellular block/flow cytometry
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard, Intracellular Block/Flow Cytometry |
| Molecular Weight (g/mol) | 18.5 |
| Name | Human IP-10 Recombinant Protein |
| Purification Method | SDS-PAGE |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction