missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Human Interleukin-1β (IL-1β)
Click to view available options
Quantity:
5 μg
Unit Size:
Each
Description
Lyophilized from a solution comprised of 0.22 ++m sterile-filtered aqueous buffered solution with no preservatives
- Reactivity: Human
- Storage Buffer: Lyophilized in an aqueous buffered solution containing BSA
- Biological Activity: The activity range is measured in a proliferation assay using TF-1 indicator cells
- Specific Activity Range: 1.0-4.0x108 Units/mg
- ED50: 25 to 100pg/ml
- Observed dose response: 100 fold
Bioassay
Specifications
Specifications
| For Use With (Application) | Bioassay |
| Formulation | Lyophilized in an aqueous Buffered solution containing BSA |
| Name | Human IL-1beta Recombinant Protein |
| Purification Method | SDS-PAGE |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Form | Lyophilized |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction