missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Human Interferon-γ inducible protein-10 (IP-10)
Click to view available options
Quantity:
10 μg
Unit Size:
Each
Description
Interferon-γ inducible protein-10 (IP-10) is a 10 kD protein that belongs to CXC chemokine family. The gene for human IP-10 has been mapped to chromosome 4q2. CXC chemokine receptor 3 (CXCR3) has been identified as the receptor for IP-10.
- Molecular weight: 10 Kda
- Purity: : The recombinant human IP-10 was produced and found to be ≥ 95% pure by SDS-PAGE analysis and visualized by silver strain
- Storage buffer: Lyophilized in an aqueous buffered solution containing BSA
Avoid multiple freeze-thaws of product.
ELISA
Specifications
Specifications
| For Use With (Application) | ELISA |
| Formulation | Lyophilized in an aqueous Buffered solution containing BSA |
| Molecular Weight (g/mol) | 10 |
| Name | Human IP-10 Recombinant Protein |
| Purification Method | SDS-PAGE |
| Quantity | 10 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Form | Lyophilized |
Research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction