missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Human IL-3
Click to view available options
Quantity:
10 μg
Unit Size:
Each
Description
- Comes as a frozen aqueous buffered solution containing glycerol, 2.0mg/mL Biotechnology grade, low endotoxin bovine serum albumin, and with no preservatives
- Can be used as a blocking control to demonstrate specificity of IL-3 staining by the directly conjugated-BVD3 1F9 antibody
Enzyme Linked Immunosorbent Assay, BioAssay
Specifications
Specifications
| Concentration | 200μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard, Intracellular Block/Flow Cytometry |
| Name | Human IL-3 Recombinant Protein |
| Quantity | 10 μg |
| Storage Requirements | -80°C |
| Recombinant | Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction