missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Human IL-15

Product Code. 15800059
Click to view available options
:
Size: 5μg; Reactivity: human
This item is not returnable. View return policy

Product Code. 15800059

Brand: BD 560254

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lyophilized sterile-filtered aqueous buffered protein solution containing 1mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives

Interleukin-15 (IL-15) is a potent immunoregulatory cytokine produced by a variety of cell types including monocytes and epithelial cells. This T cell growth factor can enhance cytolytic activity of antigen-specific T cells and lymphokine-activated killer cells. Human IL-15 is an 11 kDa protein, as determined by SDS-PAGE analysis.

  • Reactivity: Human
  • Molecular weight: 11KDa
  • >95% pure
  • Endotoxin level: ≤0.1ng per μg of human IL-15
  • Storage buffer: Lyophilized in an aqueous buffered solution containing BSA

Functional assay, ELISA Standard

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA Standard, Functional Assay
Formulation Lyophilized in an aqueous Buffered solution containing BSA
Molecular Weight (g/mol) 11
Name Human IL-15 Recombinant Protein
Quantity 5 μg
Storage Requirements -20°C
Cross Reactivity Human
Recombinant Recombinant
Form Lyophilized

Research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.