missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Erythropoietin (EPO) Protein

Product Code. 15857798 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15857798 5 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15857798 Supplier BD Supplier No. 554597
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Determined by SDS-PAGE and an absorbance assay based on Beers-Lambert law

30 kD heavily glycosylated protein containing 166 amino acids

  • Reactivity: Mouse
  • Storage Buffer: Frozen aqueous buffered solution containing BSA and glycerol
  • Measured using TF-1 indicator cells
  • Specific Activity: 0.1 - 1.0 × 108 Units/mg (Unit is defined as the amount of material required to stimulate a half-maximal response at cytokine saturation)
  • ED50: 0.1 to 1.0 ng/ml; Observed linear dose response range: >100 fold

Bioassay

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.1mg/mL
For Use With (Application) Bioassay
Formulation Frozen aqueous Buffered solution containing BSA and Glycerol
Molecular Weight (g/mol) 30
Name Mouse Erythropoietin Recombinant Protein
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Mouse
Recombinant Recombinant
Form Solution

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.