missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Voges Proskauer A Reagent Dropper
Description
To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.
- Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
- Strict quality controls assure uniformity and accuracy
Specifications
Specifications
| Type | Reagent/Stain Dropper |
| Quantity | 50 Pack |
| Certifications/Compliance | CE |
| Includes | 0.5mL of 5% w/v alpha naphthol in absolute alcohol |
| Format | Dropper |
| For Use With (Application) | Intended for use in the Voges Proskauer test |
| Storage Requirements | 15° to 30°C |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 1
- H225-Highly flammable liquid and vapour.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P233-Keep container tightly closed.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ethanol, 1-napthol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?