missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Indole Reagent Dropper
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
Description
To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.
- Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
- Strict quality controls assure uniformity and accuracy
Specifications
Specifications
| Type | Reagent/Stain Dropper |
| Quantity | 50 Pack |
| Certifications/Compliance | CE |
| Includes | One day's worth of stain |
| Format | Dropper |
| For Use With (Application) | Used in the identification of bacteria containing tryptophanase enzyme systems |
| Storage Requirements | 15° to 30°C |
| Stain Type | Indole |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P363-Wash contaminated clothing before reuse.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : isobutanol,hydrochloric acid, p-dimethylaminobenzaldehyde
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction