missing translation for 'onlineSavingsMsg'
Learn More

BD Rat Activated T Lymphocyte Cocktail

Product Code. 15839438
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15839438 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Product Code. 15839438 Supplier BD Supplier No. 558494

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

Designed to identify rat activated lymphocytes by direct immunofluorescent staining with flow cytometric analysis

  • Storage buffer: aqueous buffered solution containing BSA and ≤0.09% sodium azide
  • Components: APC anti-Rat CD3, PE anti-Rat CD25, FITC anti-Rat RT1B
  • Size: 50 tests

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Cocktail
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Quantity 50 Tests
Regulatory Status RUO
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.