missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Probetec™ ET CT/GC Specimen Collection and Dry Transport System
Click to view available options
Quantity:
100/Pk.
Description
Includes
- Male Urethral Kit: Each unit includes one rayon-tipped swab on a flexible and extendable wire in a tube
- Endocervical Kit: Each unit contains one polyurethane-tipped swab in a tube and a large rayon-tipped cleaning swab
BD ProbeTec™ ET CT/GC Amplified DNA Assay Specimen Collection and DRY TRANSPORT Kit
- Sterile, self-contained, disposable systems
- Specialized swabs for collection of urethral or endocervical specimens
- Swabs facilitate obtaining specimens and maintaining the organisms for nucleic acid amplification
- For use with the BD ProbeTec ET Chlamydia trachomatis and Neisseria gonorrhoeae Amplified DNA assays on the BD ProbeTec ET System
Endocervical Kit
- Designed for the cleaning and collection of endocervical specimens
- Large cleaning swab is included to clean the cervical os of blood and mucus prior to collection
- Polyurethane-tipped swab on a plastic shaft
- Designed for the collection of male urethral specimens
- Flexible aluminum wire and small rayon-tipped swab facilitate obtaining the specimen
- Plastic handle housing the aluminum wire and secured in the cap provides a slide action for extension of the swab
Specifications
Specifications
| Product Type | Endocervical Specimen Collection and Dry Transport Kit |
| Sterility | Sterile |
| Closure Color | Pink |
| For Use With (Equipment) | BD Probetec™ ET CT or GC amplified DNA assays |
| Includes | One Polyurethane-tipped swab in a tube and a large rayon-tipped cleaning swab |
| Quantity | 100/Pk. |
| Storage Requirements | 15°C to 30°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction