missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Pharmingen™ Hoechst 34580 Reagent
Description
- excitation: UV or violet
- emission: 368nm
- Reconstitue with 1mL distilled water to yield a 1mg/mL solution
- Ribonuclease treatment is not needed to avoid nonspecific RNA staining
Specifications
Specifications
| Content And Storage | -20°C. Protect from light. |
| For Use With (Application) | Flow Cytometry, Fluorescence microscopy |
| Quantity | 1 mg |
| Form | Lyophilized |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?