missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD PE Mouse NK Cell Separation Set - DM
For positive selection or depletion of NK and NK-T cells using BD IMagnet™
Brand: BD 558004
Additional Details : Weight : 0.02000kg
Includes: CD49b/pan-NK Cells PE, and BD IMag anti-PE particles 2
Description
- Clone: DX5
- Format: BD IMag-DM, PE
- Isotype: IgM, κ
- Reactivity: Mouse
- Immunogen: rIL-2'propagated NK1.1+ cells from C57BL/6 mice1
- Strain host: Lewis
Cell Separation
Specifications
PE | |
Rat | |
IgM κ |
Research Use Only