missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Mouse TNF ELISA Kit
Click to view available options
Quantity:
2 Plates
Unit Size:
Each
Description
- Contains solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay)
Applications:
ELISA
Specifications
Specifications
| Antigen | TNF |
| Cross Reactivity | Mouse |
| Includes | Mouse TNF standard, Mouse TNF Antibody coated plate, Enzyme concentrate Streptavidin HRP, wash concentrate (20X), stop solution, standard/Sample Diluent, Enzyme diluent, ELISA diluent II, TMB one-step Substrate reagent, detection Antibody Biotin Anti-Mouse TNF |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Quantitative determination of Mouse TNF, ELISA |
| Product Type | Mouse TNF ELISA kit |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction