Learn More
BD Mouse T Lymphocyte Subset Antibody Cocktail
Designed to identify major subsets of T lymphocytes by direct immunofluorescent staining with flow cytometric analysis
£837.08
Specifications
| Host Species | Mouse |
|---|---|
| Regulatory Status | RUO |
| Purification Method | Affinity Purified |
| Content And Storage | Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Product Code | Brand | Conjugate | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Conjugate | Price | Quantity & Availability | |||||
|
15849378
|
BD
558431 |
APC/PE/PE-Cyanine7 |
£955.00 £837.08 / Each Save £117.92 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10779856
|
BD
558391 |
FITC/PE/PE-Cyanine7 |
£870.00 £837.08 / 1 set Save £32.92 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The three antibodies have been titrated and pre-diluted, mixed together, and formulated for optimal staining performance. The Mouse T Lymphocyte Subset Isotype Control contains equivalent concentrations of fluorochrome- and isotype-matched negative-control immunoglobulin
- Permits recognition of each of three antigens on each cell in a sample
- Levels of expression of three antigens distinguish the major subpopulations of developing and peripheral T lymphocytes
- Additional fluorochrome-labeled reagents may be combined to further characterize T-cell subpopulations
- Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography
- Clone: 145-2C11, RM4-5, 53-6.7
- Isotype: Armenian Hamster IgG1, κ, Rat IgG2a, κ
Specifications
| Mouse | |
| RUO | |
| Affinity Purified | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.