missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Mouse Immunoglobulin Isotyping ELISA Kit
Click to view available options
Quantity:
80 Tests
Unit Size:
1 set
Description
Includes
8 mouse immunoglobulin isotype-specific rat monoclonal antibodies; Horseradish peroxidase (HRP)-conjugated rat anti-mouse Ig antibody; Substrate and Stop Solutions, Coating and Blocking Buffers; Positive Control Antigen Mixture
The Mouse Immunoglobulin Isotyping ELISA Kit employs direct horseradish peroxidase-labeled system and assay format eliminates the need for coating plate with antigen.
- Significant reduction in assay time without sacrificing sensitivity
ELISA
Specifications
Specifications
| Antigen | IgG |
| Cross Reactivity | Mouse |
| Includes | 1mL Rat Anti-Mouse IgG1, 1mL Rat Anti-Mouse IgG2a, 1mL Rat Anti-Mouse IgG2b, 1mL rat Anti-Mouse IgG3, 1mL Rat Anti-Mouse IgM, 1mL Rat Anti-Mouse IgA, 1mL Anti-Mouse Igkappa, 1mL rat Anti-Mouse Ig, lambda, 1mL HRP Rat Anti-Mouse Ig, 40mL Substrate reagent |
| Quantity | 80 Tests |
| Sample Type | Hybridoma Culture Supernatant |
| Sample Volume | 100 μL |
| Content And Storage | 4°C |
| For Use With (Application) | Rapid, efficient identification of Mouse immunoglobulin isotypes, ELISA |
| Product Type | Mouse Immunoglobulin isotyping ELISA kit |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction