missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Monolight™ Luminometer Thermal Printer Paper
Click to view available options
Quantity:
1 Roll
Unit Size:
1 pack
Description
- Light sensitive
- For long term storage of data, photocopying is recommended
Specifications
Specifications
| Type | Thermal Printer Paper |
| Quantity | 1 Roll |
| Content And Storage | <25°C |
| For Use With (Application) | Recording data from luminometer instrument measurements |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction