missing translation for 'onlineSavingsMsg'
Learn More

BD Lysate Treatment Reagent

Product Code. 17297454
Click to view available options
Quantity:
1 mL
Unit Size:
1 bottle
This item is not returnable. View return policy

Product Code. 17297454

Brand: BD 560309

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provided as lyophilized powder

Before use, add 1mL of pure ethanol to obtain a 50X working solution.

TRUSTED_SUSTAINABILITY

Specifications

Formulation 75.8% Phenylmethylsulfonylflouride, 4.3% Phenentroline
Quantity 1 mL
Format Powder
Product Type Lysate Treatment Reagent
Content And Storage 2° to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.