missing translation for 'onlineSavingsMsg'
Learn More

BD Jurkat Cell Lysate

Product Code. 15815939
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15815939 500 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15815939 Supplier BD Supplier No. 611451
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Convenient to use and useful control for western blotting application

  • Source is rich in endogenously expressed proteins
  • Provided in denatured format allowing for immediate loading onto SDS-PAGE gel
  • Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
  • Concentration: 1mg/mL

Routinely tested western blot

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue T-cell Leukemia
Quantity 500 μg
Storage Buffer 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Host Species Human
Concentration 1 mg/mL
Content And Storage -20°C

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.