missing translation for 'onlineSavingsMsg'
Learn More

BD Human Pluripotent Stem Cell Transcription Factor Analysis Kit

Product Code. 15830329
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15830329

missing translation for 'mfr': BD 560589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Monoclonal antibodies, isotype controls, compensation beads, staining and fixation buffers, verified protocols, and software analysis guidelines

Measuring expression of the stem cell master regulators'Nanog, Oct3/4, and Sox2'in heterogenous stem cell populations

To perform multicolor flow cytometry on human embryonic stem cells (hESCs) and induced pluripotent stem (iPS) cells

  • Reactivity: Human

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Formulation Nanog PE, Oct 3/4 PerCP-Cy™5.5, Sox2 Alexa Fluor™ 647, isotype control Antibodies, BD™ CompBead Plus compensation particles, BD Cytofix™ Fixation Buffer, BD Perm/Wash™
For Use With (Application) Flow cytometry
Includes Monoclonal Antibodies, isotype controls, compensation beads, Staining and Fixation Buffers, verified protocols and software analysis guidelines
Quantity 50 Tests
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Fixation Buffer
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : formaldehyde

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.