missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human IP-10 ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
Each
Description
Includes
Capture antibody, Detection antibody, Enzyme reagent, Standards, Instruction/analysis certificate
To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human IP-10 in serum, plasma, and cell culture supernatants.
- Flexible assay
- Calibrated against purified E.coli-expressed recombinant human IP-10
ELISA
Specifications
Specifications
| Product Type | Human IP-10 ELISA set |
| Sufficient For | 20 x 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Human IP-10 in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Human IP-10 (capture) Antibody, Biotinylated anti-Human IP-10 (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IP-10 (standard), Streptavidin-HRP |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction