missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human IL-2 sRα ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Description
Includes
Capture Antibody: Anti-human IL-2 sRα, Detection Antibody: Biotinylated anti-human IL-2 sRα, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate, Standard: Recombinant human IL-2 sRα (lyophilized), Instruction/Analysis Certificate
- Flexible assay design
- Calibrated against purified recombinant human IL-2 sRα
ELISA
Specifications
Specifications
| Concentration | 7.8 to 500pg/mL |
| Product Type | Human IL-2sRa (sCD25) ELISA set |
| Sufficient For | 20 x 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Human IL-2sRa in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Human IL-2 sRa monoclonal (capture) Antibody, Biotinylated anti-Human IL-2 sRa monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-2 sRa (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
Research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction