Learn More
BD Human Eotaxin ELISA Set
Description
Includes
- Capture Antibody: Anti-human eotaxin monoclonal antibody
- Detection Antibody: Biotinylated anti-human eotaxin monoclonal antibody
- Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standards: Recombinant human eotaxin, lyophilized
Requires
96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, Microplate reader capable of measuring absorbance at 450nm, Precision pipettes, Graduated cylinder, one liter, Deionized or distilled water, Wash bottle or automated washer, Log-log graph paper or automated data reduction, Tubes to prepare standard dilution, Laboratory timer, Plate sealers or parafilm
The OptEIA™ Set for human eotaxin contains the components necessary to develop enzyme-linked immunosorbent assays
- Standard Range: 6.3 - 400pg/mL
- Calibrated against purified Baculovirus-expressed recombinant human eotaxin
- Allow flexible assay design to fit individual laboratory needs
- To design an immunoassay with different sensitivity and dynamic range
ELISA
Specifications
Specifications
| Concentration | 6.3 to 400pg/mL |
| Product Type | Human Eotaxin ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Human eotaxin in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Human eotaxin monoclonal (capture) Antibody, Biotinylated anti-Human eotaxin monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human eotaxin (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
For research use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.