missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Rat Anti-Mouse IgD
Click to view available options
Quantity:
25 μg
50 μg
Unit Size:
Each
Beskrivelse
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Tekniske data
Tekniske data
| Product Type | BV480 Rat Anti-Mouse IgD |
| Quantity | 25 μg |
| For Use With (Application) | Flow Cytometry |
| Concentration | 0.2 mg/mL |
| Cross Reactivity | QC Testing: Mouse |
| Form | BV480 |
| Content And Storage | Store undiluted at 4°C |
| Storage Buffer | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion