missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human CD11c
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
Specifications
| Product Type | BV480 Mouse Anti-Human CD11c |
| Quantity | 25 Tests |
| For Use With (Application) | Flow Cytometry |
| Cross Reactivity | QC Testing: Human |
| Form | BV480 |
| Sample Volume | 5 μL |
| Content And Storage | Store undiluted at 4°C |
| Storage Buffer | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction