missing translation for 'onlineSavingsMsg'
Learn More

BD Fixable Viability Stain 780

Product Code. 15824289
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15824289 200 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15824289 Supplier BD Supplier No. 565388

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful for discrimination of viable from non-viable mammalian cells in multicolor flow cytometric applications. BD Horizon™ Fixable Viability Stain 780 reacts with and covalently binds to cell-surface and intracellular amines.

  • This stain is excited by the Red laser (with an excitation maximum of 759nm), and has a fluorescence emission maximum of 780nm.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 μg
For Use With (Application)
  • Flow cytometry
  • Intracellular staining (flow cytometry)
Storage Requirements -80°C
Stain Type Fixable Viability Stain 780
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.