missing translation for 'onlineSavingsMsg'
Learn More

BD HLA-A2:Ig Recombinant Soluble Dimeric Fusion Protein, Human

Product Code. 15886588
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15886588 0.05 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15886588 Supplier BD Supplier No. 551263

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Consists of three extracellular major histocompatibility complex (MHC) class I HLA-A2 domains that are fused to the VH regions of mouse IgG1, supplemented with recombinant β2M

The HLA-A2 protein was expressed together with human β2M in the mouse plasmacytoma cell line, J558L (ATCC TIB-6). The HLA-A2 and β2M polypeptide chains are associated noncovalently as a consequence of their coexpression within J558L cells. The HLA-A2:Ig fusion protein was purified from tissue culture supernatant by affinity chromatography. The purity of the preparation was confirmed by SDS-PAGE.

  • Format: Purified
  • Isotype: Mouse IgG1, λ
  • Storage buffer: Aqueous buffered solution containing ≤0.09% sodium azide
  • Purified from tissue culture supernatant by affinity chromatography
  • Purity was confirmed by SDS-PAGE

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5mg/mL
For Use With (Application) Flow Cytometry
Formulation Aqueous Buffered solution containing 0.09% Sodium Azide
Name Human HLA-A2:Ig Recombinant Fusion Protein
pH Range 7.2
Purification Method Affinity Chromatography, SDS-PAGE
Quantity 0.05 mg
Storage Requirements 4°C
Recombinant Recombinant
Form Solution

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.