missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD HeLa Cell Lysate
Description
Includes
Human Epitheloid Cervical Carcinoma
A source that can be rich in endogenously expressed proteins, this lysate is convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel.
- Storage buffer: 1mg/mL in SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
Western blot
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Epitheloid Cervical Carcinoma |
| Quantity | 500 μg |
| Storage Buffer | 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol |
| Host Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?