missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD PE h-2Kb:ig bd dimerX Recombinant Fusion Protein, Mouse
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
Description
The H-2K[b]:Ig fusion protein consists of three extracellular major histocompatibility complex (MHC) class I H-2Kb domains that are fused to VH regions of mouse IgG1.
- Format: PE
- Reactivity: Ms
- Isotype: Mouse IgG1, Λ
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | ELISA, Flow Cytometry |
| Formulation | Aqueous Buffered solution containing 0.09% Sodium Azide |
| Name | Mouse PE h-2Kb:ig bd dimerX Recombinant Fusion Protein |
| pH Range | 7.2 |
| Purification Method | Affinity Chromatography, SDS-PAGE |
| Quantity | 0.1 mg |
| Storage Requirements | 4°C |
| Recombinant | Recombinant |
| Form | Solution |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction