missing translation for 'onlineSavingsMsg'
Learn More

BD BD GolgiStop™ Protein Transport Inhibitor (containing Monensin)

Product Code. 10716676
Change view
Click to view available options
Quantity:
0.7 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10716676 0.7 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10716676 Supplier BD Supplier No. 554724
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Block the intracellular protein transport processes when added to in vitro- or in vivo-stimulated lymphoid cells

  • Produce an accumulation of cytokines and/or proteins in the Golgi complex
  • Enhance the detectability of cytokine-producing cells with immunofluorescent staining and flow cytometric analysis
Note: BD GolgiStop Protein Transport Inhibitor, containing monensin (component 51-2092KZ) contains 99.61% ethanol (w/w) and 0.26% monensin, mononatriumsalz (w/w).
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Enhance the detectability of cytokine-producing cells with immunofluorescent staining and flow cytometric analysis
pH Range 7.2
Content And Storage Store undiluted at 4°C
Product Type BD GolgiStop Protein Transport Inhibitor with Monensin
Quantity 0.7 mL
Format Liquid
compliance-icons compliance-icons
Product Identifier
  • BD GolgiStop
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ first aider if you feel unwell.
  • P330-Rinse mouth.
  • P370-In case of fire:.
  • P370+P378-In case of fire: Use to extinguish.
  • P378-Use to extinguish.
  • P403-Store in a well-ventilated place.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contain: ethanolmonensin, monosodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.