missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ EZ Gas Generating Systems and Supplies: Incubation Racks

Product Code. 11707204
Change view
Click to view available options
Type:
EZ Incubation Rack
Holds:
Petri Dishes (15 to 18)
Petri Dishes (30 to 33)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Holds unitSize
11707204 EZ Incubation Rack Petri Dishes (15 to 18) Each
11717204 EZ Incubation Rack Petri Dishes (30 to 33) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11707204 Supplier BD Supplier No. 260673
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments

Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments

  • Holds 15-18 Petri dishes
TRUSTED_SUSTAINABILITY

Specifications

Type EZ Incubation Rack
Format Box Rack
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Holds Petri Dishes (15 to 18)
Product Line GasPak
Quantity Each
Size Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.