missing translation for 'onlineSavingsMsg'
Learn More

BD FITC BrdU Flow Kit

Product Code. 10574894
Change view
Click to view available options
No. of Tests:
50
200
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
10574894 50 Each
15819098 200 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10574894 Supplier BD Supplier No. 559619

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

One 65μL vial fluorochrome-conjugated Anti-BrdU antibody, one 25mL bottle BD Cytofix/Cytoperm™ buffer, two 25mL bottles BD perm/wash™ buffer (10X), one 10mL bottle BD Cytoperm™, DNase; BrdU; 7-AAD; Kit Manual

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Includes BD Cytofix/Cytoperm™ Buffer; BD Perm/Wash™ Buffer; BD Cytoperm Plus Buffer; FITC Labeled Anti-BrdU Antibody; DNase; BrdU; 7-AAD; Kit Manual
No. of Tests 50
Regulatory Status RUO
Product Type FITC BrdU Flow Kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • FITC BrdU Flow Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H350-May cause cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: formaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.