missing translation for 'onlineSavingsMsg'
Learn More

BD ELISPOT Sets

Product Code. 15816658
Click to view available options
Antigen:
Granzyme B
IFN-gamma
IL-10
IL-2
IL-4
IL-5
IL-6
TNF
Cross Reactivity:
Human
Mouse
Product Type:
Human Granzyme B ELISPOT set
Human IFN-gamma ELISPOT set
Human IL-10 ELISPOT set
Human IL-2 ELISPOT set
Human IL-4 ELISPOT set
Human IL-5 ELISPOT set
Human TNF ELISPOT set
Mouse IFN-gamma ELISPOT set
Mouse IL-2 ELISPOT set
Mouse IL-4 ELISPOT set
Mouse IL-5 ELISPOT set
Mouse IL-6 ELISPOT set
Mouse TNF ELISPOT set
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15816658

Brand: BD 551446

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Unlabeled capture antibody (no azide/low endotoxin format), Biotinylated detection antibody, Streptavidin-HRP, 10 ELISPOT plates, Certificate of Analysis providing lot-specific optimal reagent concentrations

Sufficient capture antibody, detection antibody, and streptavidin-horseradish peroxidase for 10 x 96 wells

  • High sensitivity
  • High throughput, high content analysis
  • Minimal volume of biological material required
  • Suitable for frozen/thawed biological samples
  • Compatibility with other assays—cells analyzed by BD ELISPOT Sets can be used for cloning, proliferation assays, flow cytometry, or other methods of analysis

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Cross Reactivity Human
Includes Unlabeled capture Antibody, detection Antibody, Streptavidin HRP, 10 ELISPOT plates
Quantity 10 x 96 Tests
Content And Storage 4°C
For Use With (Application) Cloning, proliferation assays, flow cytometry or other methods of analysis
Product Type Human TNF ELISPOT set
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.