missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD DrySlide™ Reagent Slides
Description
- One-step procedure —no need for rehydrating
- No thin paper strips to manipulate and no glass slides are involved
- Each individual slide contains four reaction areas
- DrySlide Indole is a 30-second, spot indole test that uses DMABA (5% p-dimethylaminobenzaldehyde) formulation to test for indole-producing bacteria
- DrySlide Nitrocefin is a qualitative test to detect the presence of bacteria that produces β-lactamase and meets NCCLS guidelines; each slide contains a reagent cassette impregnated with nitrocefin
- DrySlide Oxidase with each slide is film-coated with an oxidase reagent (N, N, N',N'-tetramethyl-p-phenylenediamine dihydrochloride) to test for the presence of cytochrome C oxidase activity in bacteria; a positive result produces a purple color within 20 seconds
- DrySlide PYR Kit has L-pyrrolidonyl-β-naphthylamide that is used for the rapid presumptive identification of Group D Enterococci and Strep Group A; includes 15 slides and 15, 0.5mL crush ampules
Order Info
Slides are individually wrapped and supplied in convenient dispenser packs
Specifications
Specifications
| Type | Oxidase Reagent |
| Quantity | 75 Pack |
| Certifications/Compliance | CE |
| Format | Slide |
| For Use With (Application) | Used for determining the oxidase reaction of bacteria |
| Storage Requirements | 15° to 30°C |
| Stain Type | Oxidase Reagent |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?