Learn More
BD Disposable Syringes with Luer-Lok™ Tips
Product Code. 15880305
Click to view available options
Volume (Metric):
1 mL
Unit Size:
Pack of 100
Pack of 800
Product Code. 15880305
Brand: BD 309628
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
  • Sterile syringe for single use
  • Without needle
  • Clear barrel (polycarbonate or polypropylene) with bold scale markings
  • Tapered plunger rod for ease of aspiration
  • Positive plunger rod stop
  • Added BD Luer-Lok™ thread for increased secure connection
  • DNR (Latex) free
  • Shelf life: 1825 days
Specifications
Product Type Syringe
Disposable Single-use
Needle Point Number No Needle Included
Volume (Metric) 1 mL
Connection Type Luer Lock
Sterility Sterile