missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Salmonella O QC Antigens
Quick reactive Gold standard antisera-typing panels for Salmonella O antigen
£128.00 - £202.16
Specifications
Product Type | Antigen |
---|---|
For Use With (Application) | Used in the quality control of Salmonella O Antisera Poly B (225351), Factor 7 (228161), Group C1 Factors 6 (240985), and Poly A-I & Vi (240993) |
Quantity | 1 mL |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11768313
|
BD
221301 |
Antigen |
£169.00
1mL |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
11708323
|
BD
221341 |
Antigen |
£128.00
1mL |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
11798313
|
BD
221331 |
Antigen |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11778313
|
BD
221311 |
Antigen |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11788313
|
BD
221321 |
Antigen |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Product type: Antigens and Antisera
- Groupings: Q through S products
- Sub type: Quality control antigens
- Type: Antigens
- Volume: 1mL
- Shelf life: 730 days
Certifications
CE marked
Specifications
Antigen | |
Used in the quality control of Salmonella O Antisera Poly B (225351), Factor 7 (228161), Group C1 Factors 6 (240985), and Poly A-I & Vi (240993) | |
1 mL |