Learn More
BD Difco™ Fluorescent Antibody Reagents
Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light
Brand: BD 223591
Additional Details : Weight : 0.80000kg
Description
- Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light
- For use in direct fluorescent identification
Order Info
In 5mL vials.
Specifications
FA Bordetella Pertussis | |
5 mL | |
Direct fluorescent antibody technique for the identification of Bordetella pertussis and Bordetella parapertussis |